Cole-Parmer Essentials Pipette Rack, 20 to 200 µL, 10/Pack x 10; 100/CS
Representative image only.
Cole-Parmer Essentials Item # EW-15977-63
Stackable color-coded trays to house your tips in a safe and contaminate-free environment
  • Fully autoclavable and stackable
  • Color-coded tray for easy identification
  • Hinge rack allows single hand operation
  • Free from DNase, RNase, Human DNA, Pyrogen and PCR inhibitors
PRODUCT OPTIONS
$167.00USD / Case of 100
SPECS & DESCRIPTION
Specifications & Description
  • SterileNo
  • Max Volume (µL)200
  • Autoclavableyes
  • Filter TypeNone
  • Package Of100/Cs
  • Description200 uL tip rack w/o tips 10X10/Case
Key Features
  • Fully autoclavable and stackable
  • Color-coded tray for easy identification
  • Hinge rack allows single hand operation
  • Free from DNase, RNase, Human DNA, Pyrogen and PCR inhibitors
More About this Item
When it comes to reaching for that specific pipette tip, these racks make is easy. They can hold standard and long tips with various volume measurements and, are available in packages of 6 or 10 trays.