Cole-Parmer Essentials Pipette Tip, 2000 to 10000 µL, Thermo Scientific and Accumax Compatible, 100/BG x 10; 1000/CS
Representative image only.
Cole-Parmer Essentials Item # EW-15977-61
Tips free of human touch is purity at its best
  • Tips also leach-free, ensuring maximum recovery of sample yield
  • Volume measurements, along the tube are marked in increments for easy identification
  • Produced from Swiss-made Diamond Finish Mold
  • Made of FDA approved special medical grade virgin polypropylene plastic with high transparency
  • Unique label coding in packaging for easy identification
PRODUCT OPTIONS
$271.80USD / Case of 1000
$339.75USD / Case of 1000
SPECS & DESCRIPTION
Specifications & Description
  • SterileNo
  • Autoclavableyes
  • Max Volume (mL)10
  • Tip StyleStandard
  • Filter TypeNone
  • Package Of1000/Cs
  • Description10 mL Pipette Tips, 1000/Case
Key Features
  • Tips also leach-free, ensuring maximum recovery of sample yield
  • Volume measurements, along the tube are marked in increments for easy identification
  • Produced from Swiss-made Diamond Finish Mold
  • Made of FDA approved special medical grade virgin polypropylene plastic with high transparency
  • Unique label coding in packaging for easy identification
More About this Item
Not your everyday graduated pipette tips. It starts with a touch-free manufacturing facility and stringent quality control process to make these clear, plastic tips certified free of any affecting biological elements, such as human DNA, DNase and RNase and Endotoxin, or Pyrogen and PCR inhibitors. They also are noncytotoxic.
It’s about their innovative design, too. The profile and dimensions of the tips are engineered to accurately measure and transfer a volume of liquid from one container to another, as well as adapt to a wide-range of pipettes.
It ends with preserving their purity in the following packaging options, which are free of lubricants and heavy metals.
Bulk Pack: Packed in a polyethylene bag that is sealed to perfection