Cole-Parmer Essentials Replacement Sleeves, Fiberglass, Large, 2/Pk
Representative image only.
Cole-Parmer Essentials Item # EW-08126-04
Securely hold your flasks, beakers and burettes
PRODUCT OPTIONS
$12.50USD / Pkg of 2
SPECS & DESCRIPTION
Specifications & Description
  • Clamp MaterialFiberglass
  • DescriptionReplacement Sleeves, Fiberglass, Large, 2/Pk
More About this Item
Sleeves are available in both vinyl and fiberglass can be easily removed from a clamp for cleaning or replacement. Fiberglass sleeves are recommended for all applications above 212¢ªF (100¢ªC).