Cole-Parmer Essentials Replacement Sleeves, Fiberglass, Large, 3/Pk
Representative image only.
Cole-Parmer Item # EW-08126-05
Securely hold your flasks, beakers and burettes
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Clamp MaterialFiberglass
  • DescriptionReplacement Sleeves, Fiberglass, Large, 3/Pk
  • Warranty2 year
Two year warranty
Key Features
  • 2 year warranty
More About this Item
Sleeves are available in both vinyl and fiberglass can be easily removed from a clamp for cleaning or replacement. Fiberglass sleeves are recommended for all applications above 212¢ªF (100¢ªC).