Cole-Parmer Essentials UNP10404-CP Conical-Bottom Centrifuge Tubes, PP/HDPE, 50mL, Sterile; 500/CS
Representative image only.
Cole-Parmer Essentials Item # EW-78903-11
Specially designed flat threads prevent leakage
  • Polypropylene tubes are autoclavable to 250°F (121°C) Bright blue graduations are easy to read Maximum RCF range of 14,000 g DNase, RNase, pyrogen, and latex free
PRODUCT OPTIONS
$161.00USD / Case of 500
SPECS & DESCRIPTION
Specifications & Description
  • Max RCF x g14000
  • SterileYes
  • AutoclavableYes
  • Tube Capacity (mL)50
  • Tube MaterialPolyproylene
  • Tube ColorClear
  • Tube Bottom ShapeConical
  • Tube Length (mm)114.3
  • Tube Outside Diameter (OD) (mm)29.2
  • Cap IncludedYes
  • Cap StyleThreaded
  • Cap MaterialHDPE
  • GraduationsYes
  • Packaging500 per case
  • DescriptionCole-Parmer Concial Bottom Centrifuge Tubes, PP/HDPE, Sterile
Includes
HDPE screw caps.
Key Features
  • Polypropylene tubes are autoclavable to 250°F (121°C) Bright blue graduations are easy to read Maximum RCF range of 14,000 g DNase, RNase, pyrogen, and latex free
More About this Item
Available with a blue HDPE leak-proof screw cap, the clear polypropylene (PP) conical bottom centrifuge tubes feature a smooth hydrophobic internal surface to minimize wall effect. Both the tubes and caps are designed with flat threads for vacuum-type, leak-free closure at a pressure of >700 mm Hg. Bold blue graduations are easy to read, and a frosted surface area makes writing easier. Autoclavable at 121°C and 15 psi for 30 minutes. Radiation sterilized.