Cole-Parmer Ethylene Glycol Reagent Grade; 20 L

Representative image only.
Cole-Parmer Item # EW-88405-54

$342.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Empirical FormulaC2H6O2
  • Linear FormulaHOCH2CH2OH
  • ClassReagent Solvents
  • ChemistrySolvents
  • FormLiquid
  • Synonyms1,2-Ethanediol
  • Volume (mL)20000
  • Formula Weight62.07
  • PackagingCubitainer
  • DOT InformationNot regulated by DOT
  • CAS Number107-21-1
  • Shelf Life (days)60
  • DescriptionEthylene Glycol Reagent Grade, (20 L)

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat