MaterialPolypropylene bottle and cap, stainless steel vacuum pump base
BatteryNo
Power (VAC)100 to 240
Power (Hz)50/60
SterileNo
Capacity (mL)4000
DescriptionAspirating System with Hose-Barb Cap, 4 L
Warranty1 Year
Includes
pump base, 4-liter polypropylene bottle, cap with barbed couplings, tube set with filter, hand operator with single channel adapter for disposable tips, and universal power supply.
Key Features
Keep clean—all components are autoclavable except the vacuum base
Protect your vacuum pump—included filter prevents liquid aspiration into the pump
Minimize down-time when collection bottle is full—additional bottles available
Be safe—plastic bottles won't shatter if dropped
Choose from model with a hose barb couplings cap or models with quick-release couplings cap and a collection bottle level sensor
1 year warranty
More About this Item
The E-Vac aspiration system is designed to collect critical or dangerous liquids. Waste is collected in the bottle through vacuum, and pressure-sensitive button controls the right amount of vacuum for your application. All parts, except the stainless steel base containing the pump, are fully autoclavable. Two hydrophobic filters help prevent liquids from getting into the pump and pump housing in the event of an overflow.
The HandE-Vac hand operator, included with both systems options, is designed to fit the E-Vac systems or any vacuum device currently available on the market. Set vacuum by manual adjustment of the aperture control button. Ergonomic design allows for fatigue-free aspiration. The hand operator can also be used with Pasteur pipettes or multi-well plates and dishes, additional adapters required and sold separately.
Full bottles can be swapped out and autoclaved with minimum of effort and new bottles replaced within seconds. All parts, except the stainless steel base containing the pump, are fully autoclavable.