Cole-Parmer Evaporator/Concentrator for 384-well Plates; stand-alone (non-heated)
Representative image only.
Cole-Parmer Item # EW-28690-55
Indivdual needles provide equal flow of gas for faster, consistent evaporation of samples
  • Economical and reliable evaporation in a compact instrument
  • Models available to accommodate different-sized tubes and microwell plates
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Condenser TypeEvaporator
  • Total Positions384
  • DescriptionEvaporator/Concentrator for 384-well Plates; stand-alone (non-heated)
  • Warranty1 Year
1 Year Warranty
Key Features
  • Economical and reliable evaporation in a compact instrument
  • Models available to accommodate different-sized tubes and microwell plates
  • 1 year warranty
More About this Item
Rapidly evaporate common organic solvents—ideal for pharmaceutical, environmental, HPLC, GCMS, and general research applications. Needle valve controls the flow of gas into the equalizer manifold, which provides equal gas flow and uniform evaporation to each well. Needle valves ensure control for fast concentration of multiple samples. Each needle is made from 15-gauge stainless steel with easy height adjustment to accommodate a variety of sample sizes.
Stand-Alone Evaporator/Concentrators are made of autoclavable polypropylene with a nickel-plated valve; feature cutouts for easy removal of plates. Select either a 96- or 384-well model.