This item is now discontinued. We recommend the following as suggested replacements: EW-00652-30

Cole-Parmer Extended-Life Conductivity Standard, 1413 uS; 500 mL

Representative image only.
Cole-Parmer Item # EW-00651-46
Control the expiration date of your conductivity standards
  • Shelf life up to 5 years in the special storage envelope, or 1 year after opening — whichever comes first!
  • NIST-traceable calibration report from an ISO 17034 and ISO 17025 accredited laboratory included with each bottle Note: 1uS = 1Mhos
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Conductivity Value (µS)1413
  • Resistivity Value (MΩ)0.00071
  • Volume (mL)500
  • Volume (oz)16.9
  • DescriptionExtended-Life Conductivity Standard, 1413 uS; 500 mL
  • Warranty1 year
1 Year Warranty

Key Features

  • Shelf life up to 5 years in the special storage envelope, or 1 year after opening — whichever comes first!
  • NIST-traceable calibration report from an ISO 17034 and ISO 17025 accredited laboratory included with each bottle Note: 1uS = 1Mhos
  • 1 year warranty

More About this Item

Transpiration (evaporation) often limits the shelf life of conductivity standard solutions. These solutions ship in a special metal-lined envelope to extended shelf life—up to 5 years! The envelope is clearly marked with the 5-year expiration date from the date of manufacturer. When ready to use, simply open the sealed envelope and begin the 1-year shelf life. Both the bottle and Certificate of Analysis have a convenient marking area to write the new expiration date—putting you in control.
Store at room temperature. One 17-oz (500-ml) bottle. Additional values and packaging sizes are available; call for details.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat