Cole-Parmer Female Luer x 1/16" hose barb adapter, PP, 25/pk
Representative image only.
Cole-Parmer Item # EW-45508-00
Full material traceability to ensure process integrity
  • Manufactured from 100% virgin raw materials in a Class 100,000 environment
PRODUCT OPTIONS
$11.25USD / Pkg of 25
Complete the connection
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolypropylene
  • Number of Ports2
  • Connection Type-Port 1Female luer
  • Connection Type-Port 2Hose barb
  • DescriptionFemale luer x 1/16" hose barb adapter, PP, 25/pk
Certificate of Compliance
Key Features
  • Manufactured from 100% virgin raw materials in a Class 100,000 environment
More About this Item
These luer fittings are ideal for a variety of applications, including syringes or small tubing. Barbed adapters feature three styles of barbs: standard, wide-bore, and low-profile. Wide-bore barbs offer enhanced fluid flow through the fitting and are usable with any flexible tubing. Low-profile barbs are ideal for use with semi-rigid tubing, such as nylon, polyethylene, or polypropylene. Manufactured in compliance with ISO 594-1 standards, these fittings are cleaned, inspected and fully traceable. All parts meet tight dimensional tolerances for increased reliability and process purity. Choose from a range of sizes, 1/16" to 1/4", with a variety of connection options. Pack of 25.