Cole-Parmer Fine-Point Permanent Lab Markers, Black; 10/Box
Representative image only.
Cole-Parmer Item # EW-09964-00
$29.75USD / of 10
BUY MORE & SAVE
  • 1-5$29.75USD / of 10
  • 6-29$26.78USD / of 10
  • 30-119$25.29USD / of 10
  • 120+$23.80USD / of 10
SPECS & DESCRIPTION
Specifications & Description
  • ColorBlack
  • DescriptionFine-Point Permanent Lab Markers, Black; 10/Box
More About this Item
Fine-point fiber tip writes on cold, wet surfaces. Quick-drying ink permanently marks plastic; washes off glass, metal, or porcelain—won’t bleed when subjected to water or alcohol.