Sherwood Scientific Flame Photometer Automatic Dilutor
Representative image only.
Sherwood Scientific Mfr # 80600009 – Item # EW-02657-00
Accurately dilute your samples and standards
  • Maximum dilution ratio is 1:200
  • Dilution ratio drift is 0.5%
  • 1 year warranty
$7,150.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionFlame Photometer Automatic Dilutor
  • Warranty1 year
1 Year Warranty
Key Features
  • Maximum dilution ratio is 1:200
  • Dilution ratio drift is 0.5%
  • 1 year warranty
More About this Item
Automatic dilutor is designed for minimal dilutent consumption and tubing wear. Reduce blockage problems with clot catchers/constrictors. Maximum dilution ratio is 1:200 using tubing 02657-61; minimum is 1:50 using tubing 02657-62. Dilution ratio drift is 0.5% per 10 minutes.