Sherwood Scientific Flame Photometer Model 860 Autosampler
Representative image only.
Sherwood Scientific Mfr # 86000009 – Item # EW-02657-25
Provides fully automtic analysis solution
  • Idea for dual-channel and four-element flame photometers
  • Holds up to 40 vials
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Min Sample Size (µL)200
  • Sample Rate30 seconds per sample, 6 seconds in 2 second steps set by dipswitch
  • Output32 V, 1.56 A, 50 watts
  • Power (VAC)115 / 230
  • Power (Hz)50/60
  • DescriptionFlame Photometer Model 860 Autosampler
  • Warranty1 year
1 Year Warranty
Key Features
  • Idea for dual-channel and four-element flame photometers
  • Holds up to 40 vials
  • 1 year warranty
More About this Item
This autosampler provides a fully automatic analysis solution for your long sample runs - designed to work specifically with both the dual-channel and four-element flame photometers. The autosampler fits within the dimensions of the automatic dilutor (02657-00) providing the smallest footprint available, saving valuable bench space.
Operation is simple – press the “Blank” and “Calibrate” keys on the flame photometer and the probe is sent to the blank and calibrated vessels on board the autosampler carousel. The system, after calibration, is ready for the run to begin. Press the “Measure” key and walk away.
The autosampler carries the offering of the relevant sample to the automatic dilutor. Unit features automatic or manual timing settings between samples from the standard 30 seconds to up to six seconds shorter or longer. At the end of the run, defined when the last sample vial is automatically detected, the autosampler emits an audible signal to alert the operator.
Note: This autosampler requires the automatic dilutor (02657-00) for operation. Order software 02656-15 to use the Autosampler with the single-channel flame photometer.