Cole-Parmer Four Tube Frame for 150 mm Flowtube, Isolated, Aluminum
Representative image only.
Cole-Parmer Item # EW-03215-46
$319.00USD / Each
BUY MORE & SAVE
  • 1-3$319.00USD / Each
  • 4-9$287.10USD / Each
  • 10-19$271.15USD / Each
  • 20+$255.20USD / Each
Often Bought with…
SPECS & DESCRIPTION
Specifications & Description
  • Max Operating Temperature (° F)250
  • Max Operating Temperature (° C)121
  • Process Connection (NPT(F))0
  • DescriptionFour Tube Frame for 150 mm Flowtube, Isolated, Aluminum
More About this Item
These frames have valve ports which accept valve cartridges to control flow. A tripod base is required to use the multitube frame with the common inlet flow pattern; all other multitube frames are designed for panel and tripod mounting.