Cole-Parmer Freeze Spray, 10 oz; 6/Pk

Representative image only.
Cole-Parmer Item # EW-78907-01
Instantly cool electrical components to -65°F (-53.8°C)
  • Dries fast and leaves no residue
  • Quickly deliver freezing power to hard-to-reach places
  • Ultra-low global warming potential (GWP)
  • Nonflammable and ozone safe

$76.50USD / Case of 6
$115.50USD / Case of 6

SPECS & DESCRIPTION

Specifications & Description

  • FormatAerosol
  • Volume (gallons)0.08
  • Volume (Liters)0.3
  • ApplicationsLower component temperature to --65°F (-53.8°C)
  • DescriptionFreeze Spray, 10 oz; 6/Pk

Key Features

  • Dries fast and leaves no residue
  • Quickly deliver freezing power to hard-to-reach places
  • Ultra-low global warming potential (GWP)
  • Nonflammable and ozone safe

More About this Item

Cole-Parmer® Essentials freeze spray is a nonflammable aerosol that delivers freezing power exactly where needed. Instantly cool electrical components and other items to -65°F (-53.8°C). This freeze spray is ideal for lowering the temperature on electrical components to test intermittent faults, detect defective transistors, and provide protection during soldering. Saves time and eliminates water crystals caused by slower freezing methods.

Warning: Contact with skin can cause irritation or frostbite.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat