Cole-Parmer FTWORKS PC Software for Temperature Control
Representative image only.
Cole-Parmer Item # EW-16115-89
Enhance the performance of your Cole-Parmer® baths and circulators
  • Create, open, and save programs with up to 20 setpoints
  • Open, save, view and print logged data
  • Run a program in real-time mode
  • 1 year warranty
PRODUCT OPTIONS
$8.40USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithCole-Parmer® IC-300 Series immersion circulators 01262-13, 01262-15, 16101-87, 16112-50, 16112-52, 16112-53, 16112-54, 16112-55, 16112-56, and 16112-57
  • DescriptionPC Software for Temperature Control
  • Warranty1 year
1 Year Warranty
Key Features
  • Create, open, and save programs with up to 20 setpoints
  • Open, save, view and print logged data
  • Run a program in real-time mode
  • 1 year warranty
More About this Item
Lids, trays, temperature probes, and other accessories for your bath.
This easy to use control software is suitable for us with Cole-Parmer® IC-300 Series Immersion Circulators with RS-232 interface. compatible products. Use the software to specify ramp rate, hold times, unit of measure (°C or °F), and logging interval from every 5 to 60 seconds. Software make it easy to export data from the instrument for analysis. It will allow you to carry out all the tasks previously available on Calsoft and Thermsoft plus many new options on one new package.
This software has been designed to supersede the existing Calsoft and Thermsoft software packages.