This item is now discontinued. Please browse similar items
Cole-Parmer Fuse for FS-3400 and VS-4000 Centrifuges, 115 VAC
Representative image only.
Cole-Parmer Item # EW-83058-12
Fixed or variable speed for 6 or 8 place rotors
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionFuse for FS-3400 and VS-4000 Centrifuges, 115 VAC
More About this Item
Centrifuges include a fixed angle rotor to spin tubes up to 10 mL. Choose from fixed or variable speed models; both models feature a 30-minute timer, suction cup rubber feet which firmly anchor them to the benchtop, and a stainless steel guard bowl to protect against spills. A safety lid latch cuts power to the motor if the lid is raised. The exterior is made of chemically resistant plastic.
What’s included: 6 place rotor, tube shield, 6-ft power cord.