This item is now discontinued. Please browse similar items
Cole-Parmer gel-filled side-mount electrode, double-junction, 10-ft cable, BNC
Representative image only.
Cole-Parmer Item # EW-27011-10
Mount these versatile electrodes in any position
  • Durable—ideal for your demanding applications
SPECS & DESCRIPTION
Specifications & Description
  • Electrode TypeDouble junction
  • Electrode Body MaterialPVDF
  • Min pH (pH)14
  • Max Temperature (° C)100
  • Thread Size NPT(M) (in)1/2
  • ConnectorBNC
  • Cable Length (m)3
  • Descriptiongel-filled side-mount electrode, double-junction, 10-ft cable, BNC
Key Features
  • Durable—ideal for your demanding applications
More About this Item
Sealed AgCl reference cell requires no refilling during use. Rugged PVDF housing with 1/2'' NPT(M) thread withstands pressures up 100 psi, good for measurements up to 14 pH units. Includes a 10-ft cable with BNC connector.