Cole-Parmer Gel-Filled Side-Mount pH Probe, DJ/PVDF/3kOhm Balco; Spade/BNC
Representative image only.
Cole-Parmer Item # EW-27011-15
Mount these versatile electrodes in any position
  • Durable—ideal for your demanding applications
PRODUCT OPTIONS
$257.00USD / Each
BUY MORE & SAVE
  • 1-5$257.00USD / Each
  • 6-11$231.30USD / Each
  • 12-23$218.45USD / Each
  • 24+$205.60USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Electrode TypeDouble junction
  • Electrode Body MaterialPVDF
  • Min pH (pH)1
  • Max pH (pH)14
  • Max Temperature (° C)100
  • Temperature CompensationYes
  • Thread Size NPT(M) (in)1/2
  • ATC ConnectorSpade lugs
  • ATC Element3 kΩ Balco
  • ConnectorBNC
  • Cable Length (m)3
  • DescriptionGel-Filled Side-Mount pH Probe, DJ/PVDF/3kOhm Balco; Spade/BNC
Key Features
  • Durable—ideal for your demanding applications
More About this Item
Sealed AgCl reference cell requires no refilling during use. Rugged PVDF housing with 1/2'' NPT(M) thread withstands pressures up 100 psi, good for measurements up to 14 pH units. Includes a 10-ft cable with BNC connector.