Environmental Express Wide-Mouth Amber Glass Bottle, 45mm SC, 8oz, 250mL, 24/CS
Representative image only.
Environmental Express Item # EW-35205-16
Larger cap for easier sampling
  • Choose from two cleanliness levels—Level 1 include a Certificate of Analysis where Level 3 are perfect for general use
  • PTFE-lined caps provide a tight seal with excellent chemical resistance
  • Select amber glass bottles for light-sensitive samples
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionWide-Mouth Amber Glass Bottle, 45mm SC, 8oz, 250mL, 24/CS
Key Features
  • Choose from two cleanliness levels—Level 1 include a Certificate of Analysis where Level 3 are perfect for general use
  • PTFE-lined caps provide a tight seal with excellent chemical resistance
  • Select amber glass bottles for light-sensitive samples