Cole-Parmer Glass Burette, Class A, PTFE Stopcock; 50 mL

Representative image only.
Cole-Parmer Item # EW-63115-14
Positive threaded-back PTFE stopcock won't come loose during titrations
  • Class A burettes (burets) with clear borosilicate bodies and chemical-resistant PTFE stopcocks
  • White graduation markings are easy to read
  • Designed with a 50 mL capacity

PRODUCT OPTIONS

$102.10USD / Each

$62.00USD / Each

for an additional
$62.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.

SPECS & DESCRIPTION

Specifications & Description

  • Capacity (mL)50
  • Graduation Interval (mL)0.1
  • Tolerance0.05
  • DescriptionGlass Burette, Class A, PTFE Stopcock; 50 mL

Key Features

  • Class A burettes (burets) with clear borosilicate bodies and chemical-resistant PTFE stopcocks
  • White graduation markings are easy to read
  • Designed with a 50 mL capacity

More About this Item

These Class A burettes (burets) are calibrated at 20°C to exceed ASTM E287 (meets or exceeds DIN, ISO, and ASTM standards). They feature a maximum temperature of 460°F (240°C) for normal use and 750°F (400°C) for short-term use.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat