Cole-Parmer Glassware-Rescuer™ Glassware Chip File; 1/Ea
Representative image only.
Cole-Parmer Item # EW-98766-53
Save your chipped glassware!
  • Recover beakers and flasks
  • Prevent cuts and scrapes
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithGlassware
  • Length (in)8
  • Width (in)0.25
  • DescriptionGlassware-Rescuer™ Glassware Chip File; 1/Ea
1 Year Warranty
Key Features
  • Recover beakers and flasks
  • Prevent cuts and scrapes
More About this Item
Smooth out chips in any glassware with this handy industrial diamond file. Wide grip makes handling the file easy. Simple back-and-forth/brushing motion quickly refines sharp and jagged edges.