Cole-Parmer Glycerin, ACS Reagent Grade, 500 mL

Representative image only.
Cole-Parmer Item # EW-88405-63

$141.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeACS Reagent
  • ClassReagent Solvents
  • ChemistrySolvents;Analytical Reagents
  • FormLiquid
  • Volume (mL)500
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • CAS Number56-81-5
  • Shelf Life (days)60
  • DescriptionGlycerin, ACS Reagent Grade, 500 mL

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat