Cole-Parmer Hexamethylenetetramine Rgt, 500 g

Representative image only.
Cole-Parmer Item # EW-88403-76

$128.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • ClassSolids
  • ChemistrySolutions
  • FormSolid
  • Weight (g)500
  • PackagingAmber Glass Bottle
  • DOT InformationHexamethylenetetramine
  • CAS Number100-97-0
  • UN NumberUN1328
  • Shelf Life (days)60
  • DescriptionHexamethylenetetramine Rgt, 500 g

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat