Cole-Parmer High-Resolution 316 SS Metering Valve, straight flow, 6 mL/min
Representative image only.
Cole-Parmer Item # EW-03214-91
$151.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Port Size (NPT(F))0.125
  • Outlet Port1/8" NPT(F)
  • Connection TypeNPT(F)
  • Inlet Port1/8" NPT(F)
  • Orifice (in)0.042
  • Orifice (mm)1.0668
  • Max Flow Rate For Air (mL/min)200
  • Max Flow Rate For Water (mL/min)6
  • Max Temperature (° F)250
  • Max Temperature (° C)121
  • Wetted MaterialsStainless Steel
  • Flow PatternStraight
  • DescriptionHigh-Resolution 316 SS Metering Valve, straight flow, 6 mL/min
More About this Item
Brass valves feature Buna N O-rings; stainless steel valves have Viton® O-rings.