Cole-Parmer Hydrochloric Acid, 1.00N, 20 L

Representative image only.
Cole-Parmer Item # EW-88400-41
Reagents and solutions to meet your lab and research needs
  • Quality and consistency your lab can count on

PRODUCT OPTIONS

$247.25USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Normality1
  • Purity GradeComponents all ACS
  • Concentration0.0355
  • Empirical FormulaHCl
  • ClassTitrants
  • ChemistryAcids
  • FormLiquid
  • SynonymsMuriatic acid
  • Volume (mL)20000
  • Formula Weight36.46
  • PackagingCubitainer
  • DOT InformationHydrochloric Acid Solution; Class 8, Packing Group III
  • Hazard SymbolCorrosive
  • UN NumberUN1789
  • CAS Number7647-01-0
  • Shelf Life (months)24
  • DescriptionHydrochloric Acid, 1.00N, 20 L

Key Features

  • Quality and consistency your lab can count on

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat