This item is now discontinued. We recommend the following as suggested replacements: EW-14575-42
Cole-Parmer Immersion chiller with 15" flexible probe, -85°C, 120VAC
Representative image only.
Cole-Parmer Item # EW-14575-34
Temperatures as low as -85°C
  • Flexible chiller probe — shape to fit different size baths
  • 2-1/2 Years Part and Labor Warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionImmersion chiller with 15\" flexible probe, -85°C, 120VAC
  • Warranty2-1/2 Years Part and Labor
2 and a Half Year WarrantyCanadian Standards AssociationISO9001:2000 Certified Supplier
Key Features
  • Flexible chiller probe — shape to fit different size baths
  • 2-1/2 Years Part and Labor Warranty
More About this Item
Quickly cool the contents of your bath with these immersion chillers. Unit offers digital temperature control with a digital display of bath temperature and resolution of 0.1°C.
Probe can be inserted into your bath at ambient temperature — there is no need to wait until your bath reaches subzero temperatures. A generous 60" of insulated tubing (1-1⁄2'' OD) between the probe and the cabinet allows for flexibility.