Cole-Parmer Island Cabinet Assembly, 14' 6"

Representative image only.
Cole-Parmer Item # EW-33732-11
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Width (in)174
  • Height (in)36
  • Depth (in)58
  • Width (cm)441.96
  • Height (cm)91.44
  • Depth (cm)147.32
  • DescriptionIsland Cabinet Assembly, 14' 6\"
  • Warranty1 Year
1 Year Warranty

Key Features

  • 1 year warranty

More About this Item

To accommodate your lab, cabinets can serve as an island or peninsula. Each length has a 58" wide end cabinet with two doors and a 24" x 16" x 8" deep epoxy sink, and deck-mounted hot/cold 6" gooseneck faucet. Choose a reagent shelf size that best fits your needs. (Sold seperately) Cabinets with four-drawers/one-door feature one adjustable shelf each. Optional reagent shelf and phenolic pegboard are also available. Cabinets are silver gray in color with a black chemical-resistant phenolic countertop.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat