Cole-Parmer L-Glutamic Acid, poly bottle, 100 g

Representative image only.
Fisher Chemical Item # EW-88403-72
Quality and consistency your lab can count on

SPECS & DESCRIPTION

Specifications & Description

  • ClassReagent Salts
  • ChemistryAcids
  • FormSolid
  • Weight (g)100
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • CAS Number56-86-0
  • Shelf Life (days)60
  • DescriptionL-Glutamic Acid, poly bottle, 100 g

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat