Cole-Parmer Liquid Hand Sanitizer, 80% Alcohol; 1 Gallon
Representative image only.
Cole-Parmer Item # EW-78900-47
Stop the risk of cross-contamination—no rinsing required
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
  • FDA Registered (#NDC 75303-002-02)
  • Made in accordance with the World Health Organization (WHO) Formulation
  • Produced with zero nonessential additives
  • Unscented
PRODUCT OPTIONS
$39.75USD / Gallon of 1
BUY MORE & SAVE
  • 1-3$39.75USD / Gallon of 1
  • 4+$35.78USD / Gallon of 1
SPECS & DESCRIPTION
Specifications & Description
  • FormatLiquid
  • Volume (gallons)1
  • Volume (Liters)3.8
  • Dilution80% Alcohol
  • DescriptionLiquid Hand Sanitizer, 80% Alcohol; 1 Gallon
Key Features
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
  • FDA Registered (#NDC 75303-002-02)
  • Made in accordance with the World Health Organization (WHO) Formulation
  • Produced with zero nonessential additives
  • Unscented
  • Low viscosity provides a water-like consistency that is quick drying
  • Non-sticky formula
More About this Item
This liquid hand sanitizer is made following FDA/WHO Guidelines with 80% ethanol to kill 99.9% of bacteria, germs and viruses. Since no rinsing or drying is required, each application is fast and easy.
Available for sale in the continental United States only, not available for sale in Alaska or Hawaii.