Cole-Parmer Magnesium Chloride 6-Hyd, ACS Grade, 5 K

Representative image only.
Cole-Parmer Item # EW-88403-85

$270.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeACS Reagent
  • Empirical FormulaMgCl2•6H2O
  • Linear FormulaMgCl2•6H2O
  • ClassReagent Salts
  • ChemistrySolutions
  • FormSolid
  • Weight (g)5000
  • Formula Weight203.31
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • CAS Number7791-18-6
  • Shelf Life (days)60
  • DescriptionMagnesium Chloride Hexahydrate, ACS, (5 kg)

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat