Cole-Parmer Male Luer with lock ring x 1/16" hose barb, PP, 25/pk
Representative image only.
Cole-Parmer Item # EW-45518-00
Full material traceability to ensure process integrity
  • Manufactured from 100% virgin raw materials in a Class 100,000 environment
PRODUCT OPTIONS
$13.30USD / Pkg of 25
Complete the connection
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolypropylene
  • Number of Ports2
  • Connection Type-Port 1Male luer lock
  • Connection Type-Port 2Hose Barb
  • Barb Size-Port 2 (in)1/16
  • DescriptionMale luer with lock ring x 1/16" hose barb, PP, 25/pk 45518-00
Certificate of Compliance
Key Features
  • Manufactured from 100% virgin raw materials in a Class 100,000 environment
More About this Item
These luer fittings are ideal for a variety of applications, including syringes or small tubing. Barbed adapters feature three styles of barbs: standard, wide-bore, and low-profile. Wide-bore barbs offer enhanced fluid flow through the fitting and are usable with any flexible tubing. Low-profile barbs are ideal for use with semi-rigid tubing, such as nylon, polyethylene, or polypropylene. Manufactured in compliance with ISO 594-1 standards, these fittings are cleaned, inspected and fully traceable. All parts meet tight dimensional tolerances for increased reliability and process purity. Choose from a range of options including luer, thread, and hose barb.