Cole-Parmer Male Luer with Lock Ring x 1/8"ID Hose Barb, Nylon, 25/Pk
Representative image only.
Cole-Parmer Item # EW-45505-04
Cole-Parmer offers a variety of barbed luer adapters to meet your application's needs
  • Standard hose barb for use with flexible tubing
  • Smooth bore hose barb to reduce flow turbulence and risk of line contamination
  • Wide-bore hose barb for enhanced fluid path flow
  • Low-profile hose barbs for use with semi-rigid tubing such as nylon, polyethylene and polypropylene
PRODUCT OPTIONS
$10.75USD / Pkg of 25
BUY MORE & SAVE
  • 1-9$10.75USD / Pkg of 25
  • 10-49$9.68USD / Pkg of 25
  • 50-99$9.14USD / Pkg of 25
  • 100+$8.60USD / Pkg of 25
Complete the connection
SPECS & DESCRIPTION
Specifications & Description
  • MaterialNylon
  • Number of Ports2
  • Connection Type-Port 1Male luer lock
  • Barb Size-Port 2 (in)0
  • DescriptionMale luer with lock ring x 1/8" hose barb, Nylon, 25/pk
Certificate of ComplianceCole-Parmer Exclusive
Key Features
  • Standard hose barb for use with flexible tubing
  • Smooth bore hose barb to reduce flow turbulence and risk of line contamination
  • Wide-bore hose barb for enhanced fluid path flow
  • Low-profile hose barbs for use with semi-rigid tubing such as nylon, polyethylene and polypropylene
More About this Item
Both straight and elbow connectors are available. Hose barbs are available from 1/16" to 1/4" with either male or female luer connections. Male luer connections are available with either integral lock ring or separate lock ring.
REVIEWS
REVIEW SNAPSHOT®
by PowerReviews
Cole-Parmer Male luer with lock ring x 1/8" hose barb, Nylon, 25/pk
5.0
(based on 2 reviews)
Reviewed by 2 customers
Displaying reviews 1-2
Back to top
5.0
excellent fitting luer
By 
from East Lansing, MI
About Me Lab Assistant
Verified Buyer
Pros
  • Well Constructed
Cons
    Best Uses
    • In The Lab
    Comments about Cole-Parmer Male luer with lock ring x 1/8" hose barb, Nylon, 25/pk:
    Using this product to create a student lab experiment measuring pressure in sealed containers
    5.0
    C-P part better fit than OEM supplied one
    By 
    from East Lansing, MI
    About Me Lab Assistant
    Verified Buyer
    Pros
    • Functional
    Cons
      Best Uses
      • In The Lab
      Comments about Cole-Parmer Male luer with lock ring x 1/8" hose barb, Nylon, 25/pk:
      this part had a better connecting fit than OEM supplied one
      Displaying reviews 1-2
      Back to top
      Q&A
      Do you have questions about this product?
      Get answers from real customers and in-house experts with AnswerBox.
      3 Questions | 3 Answers
      Displaying questions 1-3
      • Q:
        The lock ring rotates around the conection?
        Asked on 4/12/2016 by Unai
        1 answer
        • STAFF EXPERT
          A:
          The lock ring does not rotate around the fitting, it is used to screw the female luer into and provides a tight connection.
          Answered on 4/12/2016 by Danielle from Technical Support
      • Q:
        This connector can be used to fit a female luer of 1/8''(id)x1/4''(od)?
        Asked on 11/26/2013 by Drello from Guanajuato, México
        1 answer
        • STAFF EXPERT
          A:
          This is a male luer lock on one end which fits a female luer, and a 1/8" tube ID on the other end.
          Answered on 11/27/2013 by Dan from Technical Support
      • Q:
        Can this product be autoclaved? If so, how many times can I autoclave it?
        Asked on 1/4/2013 by Tamy works fittings from CA
        1 answer
        • STAFF EXPERT
          A:
          Yes, this product can be autoclaved multiple times; though the number sterilization procedures it will tolerate is dependent on a number of application-related variables and could not be quantified.
          Answered on 1/4/2013 by Jay from Customer Service
      Displaying questions 1-3
      Do you have a question about this product? 