Cole-Parmer MCT-150-X Microcentrifuge Tube, 1.5 mL, amber, non sterile
Representative image only.
Cole-Parmer Item # EW-06333-56
General purpose microcentrifuge tubes with SnapLock cap
PRODUCT OPTIONS
$39.00USD / Pkg of 500
BUY MORE & SAVE
  • 1-5$39.00USD / Pkg of 500
  • 6-29$35.10USD / Pkg of 500
  • 30-119$33.15USD / Pkg of 500
  • 120+$31.20USD / Pkg of 500
SPECS & DESCRIPTION
Specifications & Description
  • Max RCF x g20000
  • AutoclavableYes
  • Tube Capacity (mL)1.5
  • Tube MaterialPolypropylene
  • Tube ColorAmber
  • Tube Bottom ShapeConical
  • Cap IncludedYes
  • Cap StyleClick-Fit
  • Cap MaterialPolypropylene
  • GraduationsYes
  • DescriptionMicrocentrifuge Tube, 1.5 mL, amber, non sterile
More About this Item
Cole-Parmer microcentrifuge tubes include an attached cap. All tubes feature a frosted writing surface on the tube and are graduated. Tubes are available in assorted colors (tube and cap) for quick sample identification. Boil-proof tubes are guaranteed to stay closed, even during boiling or autoclaving. All tubes are certified RNase-, DNase- and endotoxin-free for highest sample integrity