Cole-Parmer MCT-175-C-S Microcentrifuge Tube, 1.7 mL, Natural, Sterile, 250/pk
Representative image only.
Cole-Parmer Item # EW-06333-67
General purpose microcentrifuge tubes with SnapLock cap
PRODUCT OPTIONS
$23.25USD / Pkg of 250
BUY MORE & SAVE
  • 1-5$23.25USD / Pkg of 250
  • 6-29$20.93USD / Pkg of 250
  • 30-119$19.76USD / Pkg of 250
  • 120+$18.60USD / Pkg of 250
SPECS & DESCRIPTION
Specifications & Description
  • Max RCF x g20000
  • SterileYes
  • AutoclavableYes
  • Tube Capacity (mL)1.7
  • Tube MaterialPolypropylene
  • Tube ColorNatural
  • Tube Bottom ShapeAssorted
  • Cap IncludedYes
  • Cap StyleClick-Fit
  • Cap MaterialPolypropylene
  • GraduationsYes
  • DescriptionMicrocentrifuge Tube, 1.7 mL, Natural, Sterile, 250/pk
More About this Item
Cole-Parmer microcentrifuge tubes include an attached cap. All tubes feature a frosted writing surface on the tube and are graduated. Tubes are available in assorted colors (tube and cap) for quick sample identification. Boil-proof tubes are guaranteed to stay closed, even during boiling or autoclaving. All tubes are certified RNase-, DNase- and endotoxin-free for highest sample integrity