Cole-Parmer Essentials Media Bottle, PC, 500 mL, Sterile; 24/Cs

Representative image only.
Cole-Parmer Essentials Item # EW-06060-93
Safe alternative to glass
  • Use right out of the package—bottles come presterilized
  • Store tissue culture media, serum, and buffers
  • Manufactured in ISO Class 8 cleanroom
  • Comply with USP Class VI standards and are BPA-free

SPECS & DESCRIPTION

Specifications & Description

  • Capacity (mL)500
  • MaterialPolycarbonate(PC)
  • Min Graduation (mL)50
  • Max Graduation (mL)500
  • Graduation Interval (mL)50
  • Cap MaterialPolypropylene
  • Cap / Stopper SizeGL45
  • Cap ColorBlue
  • Bottle ColorTransparent
  • AutoclavableNo
  • DescriptionMedia Bottle, PC, 500 mL, Sterile; 24/Cs
Sterile

Key Features

  • Use right out of the package—bottles come presterilized
  • Store tissue culture media, serum, and buffers
  • Manufactured in ISO Class 8 cleanroom
  • Comply with USP Class VI standards and are BPA-free

More About this Item

These plastic media bottles make storing your common life science solutions simple. Gamma sterilized for convenient use—right out of the package. Molded indents and graduations make handling and reading liquid levels easy. Innovative cap design allows for easy gripping, uncapping, and stacking.
Polycarbonate (PC) bottles are autoclavable and offer good alcohol resistance. Polystyrene (PS) bottles are ideal for aqueous cell culture solutions. Gas-impermeable PETG bottles offer low-temperature stability and handle your general lab solution storage needs.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat