Cole-Parmer Membrane Service Kit with 2-mil thick PTFE Membranes
Representative image only.
Cole-Parmer Item # EW-05643-51
Get the most out of water analysis equipment
  • Replace or upgrade electrodes, probes, cells, and other accessories
PRODUCT OPTIONS
$84.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPTFE
  • Compatible WithCole-Parmer glass DO probes
  • DescriptionMembrane Service Kit with 2-mil thick PTFE Membranes
Key Features
  • Replace or upgrade electrodes, probes, cells, and other accessories
More About this Item
Water analysis accessories for Cole-Parmer® meters. Many of these items are compatible with other brands of meter.
Includes ten 2-mil thick PTFE membranes and ten silicone bands