Cole-Parmer Methanol, ACS Grade, Clear Glass Bottle, 2.5 L

Representative image only.
Cole-Parmer Item # EW-88401-44
Reagents and solutions to meet your lab and research needs
  • Quality and consistency your lab can count on

PRODUCT OPTIONS

$88.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeACS Reagent
  • Empirical FormulaCH3OH
  • ClassReagent Solvents
  • ChemistrySolvents
  • FormLiquid
  • SynonymsWood Alcohol, Methyl Alcohol
  • Volume (mL)2500
  • Formula Weight32.04
  • PackagingClear Glass Bottle
  • DOT InformationMethanol; Class 3, Packing Group II
  • Hazard SymbolFlammable
  • CAS Number67-56-1
  • UN NumberUN1230
  • Shelf Life (months)24
  • DescriptionMethanol, ACS Grade, Clear Glass Bottle, 2.5 L

Key Features

  • Quality and consistency your lab can count on

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat