Cole-Parmer Essentials Micro-90 Cleaning Solution; 4/CS
Representative image only.
Cole-Parmer Essentials Item # EW-18100-11
Removes most biological materials
  • Safe for all your ultrasonic cleaners
  • Environmentally friendly
  • Call today and ask for a sample 8-oz trial bottle of Micro-90 cleaning solution 99999-14
  • Case of four 4-Liter Bottles
  • 1 year warranty
PRODUCT OPTIONS
$315.00USD / Case of 4
SPECS & DESCRIPTION
Specifications & Description
  • FormatLiquid
  • Volume (Liters)16
  • Dilution1:100 to 2:100, 9.5 pH
  • ApplicationsManual or ultrasonic; removes blood and most biologoical materials; cleans glass, most metals, plastic, and rubber
  • DescriptionMicro-90 cleaning solution, case of four 4 Liter bottles
  • Warranty1 year
1 Year Warranty
Key Features
  • Safe for all your ultrasonic cleaners
  • Environmentally friendly
  • Call today and ask for a sample 8-oz trial bottle of Micro-90 cleaning solution 99999-14
  • Case of four 4-Liter Bottles
  • 1 year warranty
More About this Item
Micro-90 is a mild, yet powerful, free-rinsing cleaner for use in critical cleaning and general purpose applications. Solution cleans glass, most metals, plastic, rubber, fabric, and is safe for all your ultrasonic cleaners. Radioactive contaminants will not reduce cleaning capability. Solution works at any concentration and rinses clean; most applications require only 1.25 to 2.5 fluid ounces per gallon of water.
Solution is environmentally friendly—contains no phosphates, CFCs, ODCs, or borates. It safely replaces chromic acid to remove protein from glassware. Designed for use in CIP, ultrasonic and immersion applications. Can be used in validated processses. A 1% to 2% solution is effective in most applications. Typical pH: 9.5. NSF-Registered for USDA A1 use.
Note: Do not use on zinc, magnesium, or aluminum. 18100-30 shipped FOB Burlington, NJ, USA.
Do not use for private purposes (household).