Cole-Parmer Essentials Micro Aperture Stopper Green Neoprene 24pk
Representative image only.
Cole-Parmer Essentials Item # EW-62995-03
$185.00USD / Bag of 24
BUY MORE & SAVE
  • 1-5$185.00USD / Bag of 24
  • 6-29$166.50USD / Bag of 24
  • 30-119$157.25USD / Bag of 24
  • 120+$148.00USD / Bag of 24
SPECS & DESCRIPTION
Specifications & Description
  • Top Outside Diameter (OD) (mm)12.7
  • Bottom Outside Diameter (OD) (mm)3
  • Height (mm)50
  • Port Size (ID) (mm)3 to 12.7
  • ColorGreen
  • MaterialNeoprene
  • DescriptionMicro Aperture Stopper Green Neoprene 24pk
More About this Item
Use the tapered micro stoppers for openings from 4 to 12.7 mm. Available in soft, natural red rubber, or heat-resistant, green neoprene. Materials feature low sulfur content and low extractables. Cut to size.