Cole-Parmer Microscope Slide Preparation Kit
Representative image only.
Cole-Parmer Item # EW-25000-11
Convenient kit for microscopy enthusiasts and biology students
  • Kit includes clear glass slides, glass cover slips, Type A immersion oil, and a static-free cleaning cloth
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionMicroscope Slide Preparation Kit
Includes
Key Features
  • Kit includes clear glass slides, glass cover slips, Type A immersion oil, and a static-free cleaning cloth
More About this Item
Our slides feature precision ground edges on all four sides and are ideal for general purpose applications. Slides measure 75 x 25 mm and are 1 mm thick.
Cover slips are made from borosilicate glass, square (22 x 22 mm) with a thickness of #1 (0.13 to 0.17 mm).
Type A immersion oil has a viscosity of 150 cSt and a refractive index of 1.5239. Use with compound microscopes that have a 100x immersion objective.
The large (10''''W x 18''''L) lint-free cloth dissipates surface static while safely cleaning delicate surfaces — perfect for microscope objective lenses and slides. The knitted synthetic fiber is not chemically treated or wet, will not lose its effectiveness when exposed to air, and may be reused many times.