This item is now discontinued. Please browse similar items
Cole-Parmer Mid-Size System 2-Place Gel Caster Tray for 28560-00
Representative image only.
Cole-Parmer Item # EW-28560-76
Spectrum Chemical Agarose
Spectrum Chemical Agarose
$129.00 - $2,638.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionMid-Size System 2-Place Gel Caster Tray for 28560-00
More About this Item
Gel Casters allow you to precast consistently flat, even gels using the gel trays sold seperately. Casters accommodate either one or two trays. Casters feature a three-point leveling base and soft, coated neoprene side gaskets that secure trays in place and effectively seal their ends during casting — eliminating the need for taping.