Cole-Parmer Mixing Head, open bottom, anaerobic, 15 mm; 1/ea
Representative image only.
Cole-Parmer Item # EW-06064-82
Mix with low surface turbulence and high energy efficiency
  • No external blades, reduces risk of injury during cleaning
  • Made from high quality AISI 316 Stainless Steel
  • Unique design produces “full mix” with no dead spots
  • Anaerobic heads mix without air bubbles
  • Aerobic heads mix air into solutions
SPECS & DESCRIPTION
Specifications & Description
  • Material316 Stainless Steel
  • Max Volume (Liters)5
  • Width (mm)15
  • DescriptionTurbine Mixing Head, open bottom, anaerobic, 15 mm
Key Features
  • No external blades, reduces risk of injury during cleaning
  • Made from high quality AISI 316 Stainless Steel
  • Unique design produces “full mix” with no dead spots
  • Anaerobic heads mix without air bubbles
  • Aerobic heads mix air into solutions
More About this Item
These mixing heads are part of the Teras Turbin mixer assembly for uniform and homogenous mixing results inside closed containers.
Match different mixing heads, shafts, and cap assemblies to create one of four mixing styles. Order a cap mixer assembly, shaft, mixing head, and connector to create a complete mixer system.