Cole-Parmer MP(R)115 Centrifuge High Speed Fixed Angle Rotor, 6 x 50 mL
Representative image only.
Cole-Parmer Item # EW-17406-41
$1,015.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Max Speed (rpm)10000
  • Max RCF x g10600
  • Rotor TypeFixed-angle
  • Tube Sizes Accepted50 mL
  • Number Of Tubes Accepted6
  • DescriptionCentrifuge High Speed Fixed Angle Rotor, 6 x 50 mL
More About this Item
Expand and change the capacity of your centrifuge with these rotors.