This item is now discontinued. Please browse similar items
Cole-Parmer MP(R)16/MP(R)115/MP(R)315 Centrifuge Fixed Angle Rotor, 6 x 100 mL
Representative image only.
Cole-Parmer Item # EW-17406-07
SPECS & DESCRIPTION
Specifications & Description
  • Max Speed (rpm)6000
  • Min Speed (rpm)500
  • Max RCF x g4800
  • Rotor TypeFixed angle
  • Tube Sizes Accepted100 mL
  • Number Of Tubes Accepted6
  • DescriptionCentrifuge Fixed Angle Rotor, 6 x 100 mL
More About this Item
Expand and change the capacity of your centrifuge with these rotors.