Cole-Parmer MP(R)315 Centrifuge High Speed Fixed Angle Rotor, 24 x 15 mL
Representative image only.
Cole-Parmer Item # EW-17406-21
$1,429.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Max Speed (rpm)10000
  • Min Speed (rpm)500
  • Max RCF x g13400
  • Rotor TypeHigh speed fixed angle
  • Tube Sizes Accepted15 mL
  • Number Of Tubes Accepted24
  • DescriptionCentrifuge High Speed Fixed Angle Rotor, 24 x 15 mL
More About this Item
Expand and change the capacity of your centrifuge with these rotors.