Cole-Parmer MP(R)315 Centrifuge High Speed Fixed Angle Rotor, 6 x 250 mL
Representative image only.
Cole-Parmer Item # EW-17406-27
$2,701.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Max Speed (rpm)10000
  • Min Speed (rpm)500
  • Max RCF x g16100
  • Rotor TypeHigh speed fixed angle
  • Tube Sizes Accepted250 mL
  • Number Of Tubes Accepted6
  • DescriptionCentrifuge High Speed Fixed Angle Rotor, 6 x 250 mL
More About this Item
Expand and change the capacity of your centrifuge with these rotors.