This item is now discontinued. Please browse similar items
Cole-Parmer MP(R)315 Centrifuge Swingout Rotor, 4 x 750 mL
Representative image only.
Cole-Parmer Item # EW-17406-01
SPECS & DESCRIPTION
Specifications & Description
  • Max Speed (rpm)4000
  • Min RCF x g3600
  • Rotor TypeSwingout
  • Tube Sizes Accepted750 mL
  • Number Of Tubes Accepted4
  • DescriptionCentrifuge Swingout Rotor, 4 x 750 mL
More About this Item
Expand and change the capacity of your centrifuge with these rotors.