Cole-Parmer MP036 Centrifuge Fixed Angle Rotor, 6 x 50 mL
Representative image only.
Cole-Parmer Item # EW-17406-69
$532.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Max Speed (rpm)6000
  • Min Speed (rpm)500
  • Max RCF x g3800
  • Rotor TypeFixed angle
  • Tube Sizes Accepted50 mL
  • Number Of Tubes Accepted6
  • DescriptionCentrifuge Fixed Angle Rotor, 6 x 50 mL
More About this Item
Expand and change the capacity of your centrifuge with these rotors.