Cole-Parmer MP0415 Centrifuge Swingout Rotor, 8 x 10 mL
Representative image only.
Cole-Parmer Item # EW-17406-77
$395.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Max Speed (rpm)4000
  • Min Speed (rpm)500
  • Max RCF x g2200
  • Rotor TypeSwingout
  • Tube Sizes Accepted10 mL
  • Number Of Tubes Accepted8
  • DescriptionCentrifuge Swingout Rotor, 8 x 10 mL
More About this Item
Expand and change the capacity of your centrifuge with these rotors.